N-Cyclopropyl-4-nitrobenzylamine 95%

Supplier: Apollo Scientific

OR953594-1G OR953594-5G
APOSOR953594-1GEA 121 EUR
APOSOR953594-1G APOSOR953594-5G
N-Cyclopropyl-4-nitrobenzylamine 95%
N-Cyclopropyl-4-nitrobenzylamine
Formula: C₁₀H₁₂N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD07409347
CAS Number: 892579-09-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR