Ethyl [(tert-butoxycarbonyl)amino](oxo)acetate 95%

Supplier: Apollo Scientific

OR953671-5G
APOSOR953671-5GEA 213 EUR
APOSOR953671-5G
Ethyl [(tert-butoxycarbonyl)amino](oxo)acetate 95%
Ethyl [(tert-butoxycarbonyl)amino](oxo)acetate
Formula: C₉H₁₅NO₅
Storage Temperature: Freezer
MDL Number: MFCD01631201
CAS Number: 216959-34-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR