4-Methoxymethylbenzoic acid 95%

Supplier: Apollo Scientific

OR954026-1G OR954026-250MG
APOSOR954026-1GEA 487 EUR
APOSOR954026-1G APOSOR954026-250MG
4-Methoxymethylbenzoic acid 95%
4-Methoxymethylbenzoic acid
Formula: C₉H₁₀O₃
Storage Temperature: Ambient
MDL Number: MFCD06375934
CAS Number: 67003-50-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR