4-(3-Methylphenyl)phenol 96%

Supplier: Apollo Scientific

OR954768-1G OR954768-5G
APOSOR954768-1GEA 219 EUR
APOSOR954768-1G APOSOR954768-5G
4-(3-Methylphenyl)phenol 96%
4-(3-Methylphenyl)phenol
Formula: C₁₃H₁₂O
Storage Temperature: Ambient
MDL Number: MFCD01910124
CAS Number: 191724-08-0

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR