2-Formyl-4,5-dimethoxybenzonitrile 95%

Supplier: Apollo Scientific

OR955329-1G OR955329-250MG
APOSOR955329-1GEA 232 EUR
APOSOR955329-1G APOSOR955329-250MG
2-Formyl-4,5-dimethoxybenzonitrile 95%
2-Formyl-4,5-dimethoxybenzonitrile
Formula: C₁₀H₉NO₃
Storage Temperature: Ambient
MDL Number: MFCD24678322
CAS Number: 1013112-48-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR