4-{N-[2-(tert-Butyldimethylsilyloxy)ethyl]sulphamoyl}benzeneboronic acid 96%

Supplier: Apollo Scientific

OR9559-1G
APOSOR9559-1GEA 248 EUR
APOSOR9559-1G
4-{N-[2-(tert-Butyldimethylsilyloxy)ethyl]sulphamoyl}benzeneboronic acid 96%
4-{N-[2-(tert-Butyldimethylsilyloxy)ethyl]sulphamoyl}benzeneboronic acid
Formula: C₁₄H₂₆BNO₅SSi
Melting Pt: 106…108 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08436009
CAS Number: 913835-53-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR