Methyl 3-methyloxetane-3-carboxylate 95%

Supplier: Apollo Scientific

OR956028-1G
APOSOR956028-1GEA 517 EUR
APOSOR956028-1G
Methyl 3-methyloxetane-3-carboxylate 95%
Methyl 3-methyloxetane-3-carboxylate
Formula: C₆H₁₀O₃
Storage Temperature: Ambient
MDL Number: MFCD18251059
CAS Number: 1260670-18-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR