3-(N,N-Dimethylsulphamoylamino)benzeneboronic acid 96%

Supplier: Apollo Scientific
Warning

OR9571-1G OR9571-5G
APOSOR9571-1GEA 241 EUR
APOSOR9571-1G APOSOR9571-5G
3-(N,N-Dimethylsulphamoylamino)benzeneboronic acid 96%
3-(N,N-Dimethylsulphamoylamino)benzeneboronic acid
Formula: C₈H₁₃BN₂O₄S
Melting Pt: 129…132 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08235107
CAS Number: 277295-50-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR