6-(Dimethylamino)pyridin-3-ol 95%

Supplier: Apollo Scientific

OR957480-1G
APOSOR957480-1GEA 364 EUR
APOSOR957480-1G
6-(Dimethylamino)pyridin-3-ol 95%
6-(Dimethylamino)pyridin-3-ol
Formula: C₇H₁₀N₂O
Storage Temperature: Ambient
MDL Number: MFCD16998105
CAS Number: 330473-71-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR