2-Chloro-5-(propan-2-yloxy)phenol 96%

Supplier: Apollo Scientific

OR957642-1G
APOSOR957642-1GEA 364 EUR
APOSOR957642-1G
2-Chloro-5-(propan-2-yloxy)phenol 96%
2-Chloro-5-(propan-2-yloxy)phenol
Formula: C₉H₁₁ClO₂
MW: 186,635 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16997404
CAS Number: 1243374-18-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR