Ethyl 2-(4-hydroxyphenoxy)acetate 95%

Supplier: Apollo Scientific

OR957644-1G
APOSOR957644-1GEA 806 EUR
APOSOR957644-1G
Ethyl 2-(4-hydroxyphenoxy)acetate 95%
Ethyl 2-(4-hydroxyphenoxy)acetate
Formula: C₁₀H₁₂O₄
Storage Temperature: Ambient
MDL Number: MFCD12025129
CAS Number: 20872-28-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR