N,N-Diethyl-3-nitroaniline 95%

Supplier: Apollo Scientific

Synonyms: N,N-Diethyl-m-nitroaniline

OR958268-1G OR958268-250MG
APOSOR958268-1GEA 179 EUR
APOSOR958268-1G APOSOR958268-250MG
N,N-Diethyl-3-nitroaniline 95%
N,N-Diethyl-3-nitroaniline
Formula: C₁₀H₁₄N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00024459
CAS Number: 2216-16-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR