6-Methyl-2-mercaptobenzothiazole 95%

Supplier: Apollo Scientific

Synonyms: 6-Methylbenzo[d]thiazole-2-thiol

OR958424-1G
APOSOR958424-1GEA 56.7 EUR
APOSOR958424-1G
6-Methyl-2-mercaptobenzothiazole 95%
6-Methyl-2-mercaptobenzothiazole
Formula: C₈H₇NS₂
MW: 181.278 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07774732
CAS Number: 2268-79-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR