1-Bromooctane 98%

Supplier: Apollo Scientific

OR9595-1KG
APOSOR9595-1KGEA 241 EUR
APOSOR9595-1KG
1-Bromooctane 98%
1-Bromooctane
Formula: BrH₂C(CH₂)₆CH₃
Boiling Pt: 201 °C
Melting Pt: –55 °C
Density: 1,118 g/cm³ (25 °C)
Flash Pt: 80 °C
Storage Temperature: Ambient
MDL Number: MFCD00000276
CAS Number: 111-83-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR