Isoxazolo[5,4-c]pyridin-3-amine 98%

Supplier: Apollo Scientific

OR959629-1G OR959629-250MG
APOSOR959629-1GEA 178 EUR
APOSOR959629-1G APOSOR959629-250MG
Isoxazolo[5,4-c]pyridin-3-amine 98%
Isoxazolo[5,4-c]pyridin-3-amine
Formula: C₆H₅N₃O
Storage Temperature: Ambient
MDL Number: MFCD13193315
CAS Number: 114080-94-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR