2-(Cyclopropylamino)benzonitrile 97%

Supplier: Apollo Scientific

OR959901-10G OR959901-1G OR959901-5G
APOSOR959901-10GEA 754 EUR
APOSOR959901-10G APOSOR959901-1G APOSOR959901-5G
2-(Cyclopropylamino)benzonitrile 97%
2-(Cyclopropylamino)benzonitrile
Formula: C₁₀H₁₀N₂
Storage Temperature: Ambient
MDL Number: MFCD11155689
CAS Number: 675575-45-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR