3-(2-Methoxyphenoxy)propionic acid 95%

Supplier: Apollo Scientific

OR962573-1G OR962573-5G
APOSOR962573-1GEA 143 EUR
APOSOR962573-1G APOSOR962573-5G
3-(2-Methoxyphenoxy)propionic acid 95%
3-(2-Methoxyphenoxy)propionic acid
Formula: C₁₀H₁₂O₄
Storage Temperature: Ambient
MDL Number: MFCD01931914
CAS Number: 20370-80-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR