Glycyl-L-leucine 98%

Supplier: Apollo Scientific
Warning

OR963728-100G OR963728-25G
APOSOR963728-100GEA 430 EUR
APOSOR963728-100G APOSOR963728-25G
Glycyl-L-leucine 98%
Glycyl-L-leucine
Formula: C₈H₁₆N₂O₃
Storage Temperature: Ambient
MDL Number: MFCD00008127
CAS Number: 869-19-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR