H-Pro-OEt hydrochloride 97%

Supplier: Apollo Scientific

OR963811-25G
APOSOR963811-25GEA 125 EUR
APOSOR963811-25G
H-Pro-OEt hydrochloride 97%
H-Pro-OEt hydrochloride
Formula: C₇H₁₃NO₂·ClH
Storage Temperature: Ambient
MDL Number: MFCD00191004
CAS Number: 33305-75-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR