3-Butyn-1-amine hydrochloride 95%

Supplier: Apollo Scientific
Warning

OR964735-250MG OR964735-5G OR964735-25G
APOSOR964735-250MGEA 0 EUR
APOSOR964735-250MG APOSOR964735-5G APOSOR964735-25G
3-Butyn-1-amine hydrochloride 95%
3-Butyn-1-amine hydrochloride
Formula: C₄H₇N·HCl
Storage Temperature: Ambient
MDL Number: MFCD06658391
CAS Number: 88211-50-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR