1-sec-Butyl-5-oxopyrrolidine-3-carboxylic acid 95%

Supplier: Apollo Scientific

OR965024-1G OR965024-5G
APOSOR965024-1GEA 190 EUR
APOSOR965024-1G APOSOR965024-5G
1-sec-Butyl-5-oxopyrrolidine-3-carboxylic acid 95%
1-sec-Butyl-5-oxopyrrolidine-3-carboxylic acid
Formula: C₉H₁₅NO₃
Storage Temperature: Ambient
MDL Number: MFCD03834530
CAS Number: 696647-92-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR