1-(1-Methyl-1H-pyrazol-5-yl)ethanone 95%

Supplier: Apollo Scientific

OR965394-1G OR965394-250MG
APOSOR965394-1GEA 538 EUR
APOSOR965394-1G APOSOR965394-250MG
1-(1-Methyl-1H-pyrazol-5-yl)ethanone 95%
1-(1-Methyl-1H-pyrazol-5-yl)ethanone
Formula: C₆H₈N₂O
MW: 124.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04968784
CAS Number: 137890-05-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR