2-(3-Chlorophenyl)ethanol 98%

Supplier: Apollo Scientific

OR965862-100G OR965862-25G
APOSOR965862-100GEA 213 EUR
APOSOR965862-100G APOSOR965862-25G
2-(3-Chlorophenyl)ethanol 98%
2-(3-Chlorophenyl)ethanol
Formula: C₈H₉ClO
Storage Temperature: Ambient
MDL Number: MFCD00002892
CAS Number: 5182-44-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR