1-(4-Bromophenyl)-2-pyrrolidinone

Supplier: Apollo Scientific
Warning

Synonyms: 1-(4-Bromophenyl)pyrrolidin-2-one

OR9934-1G OR9934-5G OR9934-250MG
APOSOR9934-1GEA 72.5 EUR
APOSOR9934-1G APOSOR9934-5G APOSOR9934-250MG
1-(4-Bromophenyl)-2-pyrrolidinone
1-(4-Bromophenyl)-2-pyrrolidinone
Formula: C₁₀H₁₀BrNO
Melting Pt: 98…101 °C
Storage Temperature: Ambient
MDL Number: MFCD00138502
CAS Number: 7661-32-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR