L-Cysteine-glutathione disulphide

Supplier: Apollo Scientific
Warning

Synonyms: (2S)-2-Amino-5-({(2R)-3-{[(2R)-2-amino-2-carboxyethyl]disulphanyl}-1-[(carboxymethyl)amino]-1-oxoprop-2-yl}amino)-5-oxpentanoic acid

OR9955T-10MG
APOSOR9955T-10MGEA 451 EUR
APOSOR9955T-10MG
L-Cysteine-glutathione disulphide
L-Cysteine-glutathione disulphide
Formula: C₁₃H₂₂N₄O₈S₂
Melting Pt: 234 °C (dec.)
Storage Temperature: Freezer
MDL Number: MFCD00269957
CAS Number: 13081-14-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR