2,2,3,3,3-Pentafluoropropyl-p-toluenesulphonate 98%

Supplier: Apollo Scientific
Warning

Synonyms: p-Toluenesulphonic acid 2,2,3,3,3-Pentafluoropropyl Ester, 2,2,3,3,3-Pentafluoropropyl Tosylate, 2,2,3,3,3-Pentafluoropropyl 4-methylbenzenesulphonate

PC0022-100G PC0022-25G
APOSPC0022-100GEA 676 EUR
APOSPC0022-100G APOSPC0022-25G
2,2,3,3,3-Pentafluoropropyl-p-toluenesulphonate 98%
2,2,3,3,3-Pentafluoropropyl-p-toluenesulphonate
Formula: C₁₀H₉F₅O₃S
MW: 304.23 g/mol
Boiling Pt: 75 °C/0.75mmHg
Melting Pt: 53…55 °C
Storage Temperature: Ambient
MDL Number: MFCD02093346
CAS Number: 565-42-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR