2-Fluoro-3-hydroxypyridine

Supplier: Apollo Scientific
Warning

PC01476-25G PC01476-5G PC01476-100G
APOSPC01476-25GEA 58 EUR
APOSPC01476-25G APOSPC01476-5G APOSPC01476-100G
2-Fluoro-3-hydroxypyridine
2-Fluoro-3-hydroxypyridine
Formula: C₅H₄FNO
Melting Pt: 131…133 °C
Storage Temperature: Ambient
MDL Number: MFCD04112569
CAS Number: 174669-74-0

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR