3-Iodo-4-(trifluoromethoxy)nitrobenzene

Supplier: Apollo Scientific
Warning

PC01489-1G PC01489-5G PC01489-25G
APOSPC01489-1GEA 69.6 EUR
APOSPC01489-1G APOSPC01489-5G APOSPC01489-25G
3-Iodo-4-(trifluoromethoxy)nitrobenzene
3-Iodo-4-(trifluoromethoxy)nitrobenzene
Formula: C₇H₃F₃INO₃
Storage Temperature: Ambient
MDL Number: MFCD13194454
CAS Number: 194344-29-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR