3-Fluoro-4-(trifluoromethyl)benzyl alcohol 97%

Supplier: Apollo Scientific
Warning

PC0304-1G PC0304-5G PC0304-25G
APOSPC0304-1GEA 21.9 EUR
APOSPC0304-1G APOSPC0304-5G APOSPC0304-25G
3-Fluoro-4-(trifluoromethyl)benzyl alcohol 97%
3-Fluoro-4-(trifluoromethyl)benzyl alcohol
Formula: C₈H₆F₄O
Storage Temperature: Ambient
MDL Number: MFCD00236285
CAS Number: 230295-16-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR