3-Fluoro-4-methylbenzyl alcohol

Supplier: Apollo Scientific
Warning

PC0351-10G PC0351-1G PC0351-5G PC0351-25G
APOSPC0351-10GEA 159 EUR
APOSPC0351-10G APOSPC0351-1G APOSPC0351-5G APOSPC0351-25G
3-Fluoro-4-methylbenzyl alcohol
3-Fluoro-4-methylbenzyl alcohol
Formula: C₈H₉FO
Melting Pt: 33…36 °C
Storage Temperature: Ambient
MDL Number: MFCD01631533
CAS Number: 192702-79-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR