4-Hydroxy-2-(trifluoromethyl)benzoic acid 97%

Supplier: Apollo Scientific
Warning

PC0398-1G
APOSPC0398-1GEA 102 EUR
APOSPC0398-1G
4-Hydroxy-2-(trifluoromethyl)benzoic acid 97%
4-Hydroxy-2-(trifluoromethyl)benzoic acid
Formula: C₈H₅F₃O₃
Melting Pt: 152,5…161 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD01091007
CAS Number: 320-32-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR