4-Methoxy-3-(trifluoromethyl)benzonitrile

Supplier: Apollo Scientific
Warning

PC0400-25G PC0400-5G PC0400-100G
APOSPC0400-25GEA 145 EUR
APOSPC0400-25G APOSPC0400-5G APOSPC0400-100G
4-Methoxy-3-(trifluoromethyl)benzonitrile
4-Methoxy-3-(trifluoromethyl)benzonitrile
Formula: C₉H₆F₃NO
Melting Pt: 98…100 °C
Storage Temperature: Ambient
MDL Number: MFCD01631575
CAS Number: 261951-87-5
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR