α-(Trifluoromethyl)styrene 97%

Supplier: Apollo Scientific
Warning

Synonyms: [1-(Trifluoromethyl)vinyl]benzene, (3,3,3-Trifluoroprop-1-en-2-yl)benzene, Alpha-(trifluoromethyl)styrene

PC0401-1G
APOSPC0401-1GEA 250 EUR
APOSPC0401-1G
α-(Trifluoromethyl)styrene 97%
α-(Trifluoromethyl)styrene
Formula: C₉H₇F₃
MW: 172.147 g/mol
Boiling Pt: 148…148.5 °C
Density: 1.167 g/cm³ (25 °C)
Flash Pt: 37 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00081197
CAS Number: 384-64-5
UN: 1993
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR