2-(Trifluoromethoxy)anisole 97%

Supplier: Apollo Scientific
Warning

Synonyms: 1-Methoxy-2-(trifluoromethoxy)benzene, 2-Trifluoromethoxyanisole

PC0515-1G PC0515-5G PC0515-25G
APOSPC0515-1GEA 21.9 EUR
APOSPC0515-1G APOSPC0515-5G APOSPC0515-25G
2-(Trifluoromethoxy)anisole 97%
2-(Trifluoromethoxy)anisole
Formula: C₈H₇F₃O₂
MW: 192.14 g/mol
Boiling Pt: 69…70 °C/10mmHg
Flash Pt: 47 °C
Storage Temperature: Ambient
CAS Number: 261952-22-1
UN: 1993
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR