3-(4-Fluorophenoxy)benzyl bromide 97%

Supplier: Apollo Scientific

PC0853-1G PC0853-5G
APOSPC0853-1GEA 328 EUR
APOSPC0853-1G APOSPC0853-5G
3-(4-Fluorophenoxy)benzyl bromide 97%
3-(4-Fluorophenoxy)benzyl bromide
Formula: C₁₃H₁₀BrFO
Boiling Pt: 160 °C/2mmHg
Storage Temperature: Refrigerator
MDL Number: MFCD00059915
CAS Number: 65295-58-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR