Methyl-2-fluoro-6-methoxybenzoate

Supplier: Apollo Scientific
Warning

PC0870-1G PC0870-5G PC0870-25G
APOSPC0870-1GEA 21.9 EUR
APOSPC0870-1G APOSPC0870-5G APOSPC0870-25G
Methyl-2-fluoro-6-methoxybenzoate
Methyl 2-fluoro-6-methoxybenzoate
Formula: C₉H₉FO₃
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD03788537
CAS Number: 178747-79-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR