2'-Chloro-6'-fluoroacetophenone 97%

Supplier: Apollo Scientific
Warning

PC0902-250MG PC0902-25G PC0902-5G PC0902-100G
APOSPC0902-250MGEA 21.9 EUR
APOSPC0902-250MG APOSPC0902-25G APOSPC0902-5G APOSPC0902-100G
2'-Chloro-6'-fluoroacetophenone 97%
2'-Chloro-6'-fluoroacetophenone
Formula: C₈H₆ClFO
Storage Temperature: Ambient
MDL Number: MFCD01631390
CAS Number: 87327-69-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR