3,4,4-Trifluorobut-3-en-1-ol 97%

Supplier: Apollo Scientific
Warning

Synonyms: 1,1,2-Trifluorobut-1-en-4-ol, 4-Hydroxy-1,1,2-trifluorobut-1-ene

PC0990-5G
APOSPC0990-5GEA 473 EUR
APOSPC0990-5G
3,4,4-Trifluorobut-3-en-1-ol 97%
3,4,4-Trifluorobut-3-en-1-ol
Formula: C₄H₅F₃O
MW: 126.077 g/mol
Boiling Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD07784254
CAS Number: 97168-13-3
UN: 1993
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR