2-[3-(Trifluoromethyl)phenoxy]nicotinic acid ≥95%

Supplier: Apollo Scientific
Warning

PC10175-1G
APOSPC10175-1GEA 270 EUR
APOSPC10175-1G
2-[3-(Trifluoromethyl)phenoxy]nicotinic acid ≥95%
2-[3-(Trifluoromethyl)phenoxy]nicotinic acid
Formula: C₁₃H₈F₃NO₃
Melting Pt: 156…159 °C
Storage Temperature: Ambient
MDL Number: MFCD01938784
CAS Number: 36701-89-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR