4-(4-Fluorophenyl)-1H-pyrazol-5-amine

Supplier: Apollo Scientific
Warning

PC10192-1G
APOSPC10192-1GEA 109 EUR
APOSPC10192-1G
4-(4-Fluorophenyl)-1H-pyrazol-5-amine
4-(4-Fluorophenyl)-1H-pyrazol-5-amine
Formula: C₉H₈FN₃
MW: 177.18 g/mol
Melting Pt: 174…175 °C
Storage Temperature: Ambient
MDL Number: MFCD03001202
CAS Number: 5848-04-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR