3-Amino-3-[3-(trifluoromethyl)phenyl]propanoic acid

Supplier: Apollo Scientific

PC10262-1G
APOSPC10262-1GEA 89.9 EUR
APOSPC10262-1G
3-Amino-3-[3-(trifluoromethyl)phenyl]propanoic acid
3-Amino-3-[3-(trifluoromethyl)phenyl]propanoic acid
Formula: C10H10F3NO2
Melting Pt: 210…212 °C
Storage Temperature: Ambient
MDL Number: MFCD02653329
CAS Number: 143438-91-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR