4-Bromo-3-fluoro-2-nitroaniline 97%

Supplier: Apollo Scientific
Warning

PC10401-1G PC10401-5G PC10401-10G
APOSPC10401-1GEA 23.2 EUR
APOSPC10401-1G APOSPC10401-5G APOSPC10401-10G
4-Bromo-3-fluoro-2-nitroaniline 97%
4-Bromo-3-fluoro-2-nitroaniline
Formula: C₆H₄BrFN₂O₂
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07368682
CAS Number: 886762-75-0
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR