2-Fluoro-5-iodopyridine 98%

Supplier: Apollo Scientific
Warning

PC10421-100G PC10421-25G PC10421-5G
APOSPC10421-100GEA 275 EUR
APOSPC10421-100G APOSPC10421-25G APOSPC10421-5G
2-Fluoro-5-iodopyridine 98%
2-Fluoro-5-iodopyridine
Formula: C₅H₃FIN
Melting Pt: 35…36 °C
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD03095301
CAS Number: 171197-80-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR