2-Bromo-5-fluoro-6-methylpyridine 98%

Supplier: Apollo Scientific
Warning

PC10422-25G PC10422-5G PC10422-10G
APOSPC10422-25GEA 78.3 EUR
APOSPC10422-25G APOSPC10422-5G APOSPC10422-10G
2-Bromo-5-fluoro-6-methylpyridine 98%
2-Bromo-5-fluoro-6-methylpyridine
Formula: C₆H₅BrFN
Melting Pt: 63…65 °C
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD03095064
CAS Number: 374633-38-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR