2-Fluoro-6-(trifluoromethyl)aniline 98%

Supplier: Apollo Scientific
Warning

PC1071R-1G PC1071R-5G PC1071R-25G
APOSPC1071R-1GEA 21.9 EUR
APOSPC1071R-1G APOSPC1071R-5G APOSPC1071R-25G
2-Fluoro-6-(trifluoromethyl)aniline 98%
2-Fluoro-6-(trifluoromethyl)aniline
Formula: C₇H₅F₄N
Boiling Pt: 155 °C
Density: 1,388 g/cm³ (25 °C)
Flash Pt: 53 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00061242
CAS Number: 144851-61-6
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR