Ammonium hydrogen difluoride 98%

Supplier: Apollo Scientific

Synonyms: Ammonium bifluoride, Ammonium hydrogen fluoride

PC1104-500G
APOSPC1104-500GEA 104 EUR
APOSPC1104-500G
Ammonium hydrogen difluoride 98%
Ammonium hydrogen difluoride
Formula: NH₄HF₂
Boiling Pt: 230 °C/760mmHg
Melting Pt: 127 °C
Density: 1,5 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012018
CAS Number: 1341-49-7
UN: 1727
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR