1H,1H-Heptafluoro-n-butyl methacrylate 97%

Supplier: Apollo Scientific
Warning

PC11102-10G PC11102-25G
APOSPC11102-10GEA 118 EUR
APOSPC11102-10G APOSPC11102-25G
1H,1H-Heptafluoro-n-butyl methacrylate 97%
1H,1H-Heptafluoro-n-butyl methacrylate
Formula: C₈H₇F₇O₂
Boiling Pt: 134…136 °C
Density: 1,345 g/cm³ (25 °C)
Flash Pt: 36 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00039251
CAS Number: 13695-31-3
EINECS: 237-217-7
UN: 3272
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR