trans-1,2-Bis(perfluoro-n-butyl)ethylene 99%

Supplier: Apollo Scientific
Warning

Synonyms: (5E)-5H,6H-Octadecafluorodec-5-ene

PC1237-5G
APOSPC1237-5GEA 271 EUR
APOSPC1237-5G
trans-1,2-Bis(perfluoro-n-butyl)ethylene 99%
trans-1,2-Bis(perfluoro-n-butyl)ethylene
Formula: C₁₀H₂F₁₈
MW: 464.1 g/mol
Boiling Pt: 64 °C/56mmHg
Density: 1.675 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00055550
CAS Number: 84551-43-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR