2-Nitro-4-(trifluoromethyl)phenylhydrazine hydrochloride

Supplier: Apollo Scientific
Warning

PC1277-1G PC1277-25G PC1277-5G
APOSPC1277-1GEA 140 EUR
APOSPC1277-1G APOSPC1277-25G APOSPC1277-5G
2-Nitro-4-(trifluoromethyl)phenylhydrazine hydrochloride
2-Nitro-4-(trifluoromethyl)phenylhydrazine hydrochloride
Formula: C₇H₆F₃N₃O₂·ClH
Storage Temperature: Ambient
MDL Number: MFCD04966802
CAS Number: 1049748-69-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR