4-Chloro-2-fluorobenzotrifluoride

Supplier: Apollo Scientific
Warning

Synonyms: 4-Chloro-2-fluoro-1-(trifluoromethyl)benzene

PC1388-1G
APOSPC1388-1GEA 21.9 EUR
APOSPC1388-1G
4-Chloro-2-fluorobenzotrifluoride
4-Chloro-2-fluorobenzotrifluoride
Formula: C₇H₃ClF₄
MW: 198,5453 g/mol
Density: 1,5 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD03412199
CAS Number: 94444-59-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR